APO455830693 Apollo Scientific Ltd

5-Methyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD06738813
  • Cas Number : 120237-76-5
  • Molecular Weight : 143.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-1,3-thiazole-4-carboxylic acid
Inchi Code 1S/C5H5NO2S/c1-3-4(5(7)8)6-2-9-3/h2H,1H3,(H,7,8)
Inchi Key OIWNDJQQIMISPC-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-5-methyl-1,3-thiazole
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methyl-13-thiazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-13-thiazole-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart