APO455830695 Apollo Scientific Ltd

(2S)-2-(Aminomethyl)-4,4-diethoxypiperidine fumarate, 2-BOC protected

  • MDL number MFCD09953464
  • Cas Number : 1301706-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl [(2S)-4,4-diethoxypiperidin-2-ylmethyl]carbamate (2E)-but-2-enedioate
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=45357

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=45357
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart