APO455830697 Apollo Scientific Ltd

6-Bromo-3-chloroisoquinoline

  • MDL number MFCD08703290
  • Cas Number : 552331-06-3
  • Molecular Weight : 242.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-chloroisoquinoline
Inchi Code 1S/C9H5BrClN/c10-8-2-1-6-5-12-9(11)4-7(6)3-8/h1-5H
Inchi Key HYCCUWZOOADPGN-UHFFFAOYSA-N

Description

Synonym 6-Bromo-3-chloro-2-azanaphthalene
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-3-chloroisoquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-3-chloroisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart