APO455830703 Apollo Scientific Ltd

7-Bromo-3-chloroisoquinoline

  • MDL number MFCD11044699
  • Cas Number : 1029720-65-7
  • Molecular Weight : 242.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromo-3-chloroisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromo-3-chloroisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart