APO455830705 Apollo Scientific Ltd

2-Chloropyrimidine-5-carboxylic acid

  • MDL number MFCD06739036
  • Cas Number : 374068-01-6
  • Molecular Weight : 158.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Carboxy-2-chloropyrimidine, 5-Carboxy-2-chloro-1,3-diazine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloropyrimidine-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloropyrimidine-5-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart