APO455830711 Apollo Scientific Ltd

Isoquinoline-3-carboxaldehyde

  • MDL number MFCD06227437
  • Cas Number : 5470-80-4
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Formylisoquinoline, 3-Formyl-2-azanaphthalene
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/isoquinoline-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/isoquinoline-3-carboxaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart