APO455830715 Apollo Scientific Ltd

[5-(2-Methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine dihydrochloride

  • MDL number MFCD07704030
  • Cas Number : 1030012-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Aminomethyl)-5-(2-methyl-1,3-thiazol-4-yl)thiophene dihydrochloride, 4-[5-(Aminomethyl)thien-2-yl]-2-methyl-1,3-thiazole dihydrochloride
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47824

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47824
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart