APO455830718 Apollo Scientific Ltd

4-Amino-3-methylbenzenesulphonic acid

  • MDL number MFCD00025198
  • Cas Number : 98-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 2-Amino-5-sulphotoluene, 2-Methyl-4-sulphoaniline
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47042

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47042
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart