APO455830725 Apollo Scientific Ltd

3-[Di(but-1-yl)amino]propan-1-ol

  • MDL number MFCD00152393
  • Cas Number : 2050-51-3
  • Molecular Weight : 187.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(dibutylamino)-1-propanol
Inchi Code 1S/C11H25NO/c1-3-5-8-12(9-6-4-2)10-7-11-13/h13H,3-11H2,1-2H3
Inchi Key DMPODMBXLRMZSP-UHFFFAOYSA-N

Description

Synonym N-Butyl-N-(3-hydroxyprop-1-yl)butan-1-amine
Flash Point 94-97/3mm
Storage temp. Store at 2-8
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-dibut-1-ylaminopropan-1-ol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-dibut-1-ylaminopropan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart