APO455830727 Apollo Scientific Ltd

3-Amino-4-butoxybenzoic acid

  • MDL number MFCD12168705
  • Cas Number : 855202-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Butoxy-5-carboxyaniline
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47857

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47857
pictograms GHS07
hstatements H302;H312;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart