APO455830733 Apollo Scientific Ltd

N-(2,3-Dimethylbenzyl)cyclopropylamine hydrochloride

  • MDL number MFCD17168270
  • Cas Number : 909702-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Cyclopropyl-2,3-dimethylbenzylamine hydrochloride, 3-[(Cyclopropylamino)methyl]-o-xylene hydrochloride
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47869

Safety Information

signalword Danger
pcodes P280;P271;P309+P311
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47869
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart