APO455830747 Apollo Scientific Ltd

1-(5-Methyl-3-phenylisoxazol-4-yl)ethan-1-ol

  • MDL number MFCD16631269
  • Cas Number : 6497-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-104
Synonym 1-(5-Methyl-3-phenyl-1,2-oxazol-4-yl)ethan-1-ol, 4-(1-Hydroxyethyl)-5-methyl-3-phenylisoxazole
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48090

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48090
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart