APO455830748 Apollo Scientific Ltd

Ethyl 2-chloropyrimidine-5-carboxylate

  • MDL number MFCD09863164
  • Cas Number : 89793-12-4
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-chloro-5-pyrimidinecarboxylate
Inchi Code 1S/C7H7ClN2O2/c1-2-12-6(11)5-3-9-7(8)10-4-5/h3-4H,2H2,1H3
Inchi Key IEMKQRSOAOPKRJ-UHFFFAOYSA-N

Description

Synonym 2-Chloro-5-(ethoxycarbonyl)pyrimidine, 2-Chloro-5-(ethoxycarbonyl)-1,3-diazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-chloropyrimidine-5-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-chloropyrimidine-5-carboxylate
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart