APO455830750 Apollo Scientific Ltd

3-Bromo-5-methoxybenzaldehyde

  • MDL number MFCD06797975
  • Cas Number : 262450-65-7
  • Molecular Weight : 215.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 68-72
Iupac Name 3-bromo-5-methoxybenzaldehyde
Inchi Code 1S/C8H7BrO2/c1-11-8-3-6(5-10)2-7(9)4-8/h2-5H,1H3
Inchi Key GCXBEEKTPWZHJN-UHFFFAOYSA-N

Description

Synonym 3-Bromo-5-formylanisole, 5-Bromo-m-anisaldehyde
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 68-72
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-methoxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart