APO455830759 Apollo Scientific Ltd

4-(Bromomethyl)-3-iodobenzonitrile

  • MDL number MFCD16872046
  • Cas Number : 182287-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Cyano-2-iodobenzyl bromide
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47667

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47667
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart