APO455830762 Apollo Scientific Ltd

2-(Aminomethyl)-4-methylpyridine hydrochloride

  • MDL number MFCD16628187
  • Cas Number : 1257535-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (4-Methylpyridin-2-yl)methylamine hydrochloride, 4-Methylpicolinylamine hydrochloride
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47593

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47593
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart