APO455830781 Apollo Scientific Ltd

3-Bromo-4-iodophenol

  • MDL number MFCD17000017
  • Cas Number : 1037298-05-7
  • Molecular Weight : 298.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-4-iodophenol
Inchi Code 1S/C6H4BrIO/c7-5-3-4(9)1-2-6(5)8/h1-3,9H
Inchi Key GHYOKMYZWDAXJW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-iodophenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-iodophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart