APO455830787 Apollo Scientific Ltd

4-Bromo-3-nitroaniline

  • MDL number MFCD01320678
  • Cas Number : 53324-38-2
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-3-nitroaniline
Inchi Code 1S/C6H5BrN2O2/c7-5-2-1-4(8)3-6(5)9(10)11/h1-3H,8H2
Inchi Key PITHQPMZWKZGRZ-UHFFFAOYSA-N

Description

Synonym 5-Amino-2-bromonitrobenzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart