APO455830789 Apollo Scientific Ltd

2-Amino-4-chloro-3-nitropyridine

  • MDL number MFCD04116097
  • Cas Number : 6980-08-1
  • Molecular Weight : 173.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-3-nitropyridin-2-amine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-chloro-3-nitropyridine

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-chloro-3-nitropyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart