APO455830793 Apollo Scientific Ltd

Bis(4-phenoxyphenyl)methanone

  • MDL number MFCD00077969
  • Cas Number : 14984-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-151
Synonym 4,4'-Diphenoxybenzophenone
Purity 99
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=49010

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=49010
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart