APO455830797 Apollo Scientific Ltd

3-Amino-6-bromopyrazine-2-carboxylic acid

  • MDL number MFCD08275681
  • Cas Number : 486424-37-7
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Synonym 2-Amino-5-bromo-3-carboxypyrazine, 2-Amino-5-bromo-3-carboxy-1,4-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-6-bromopyrazine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-6-bromopyrazine-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart