APO455830798 Apollo Scientific Ltd

Methyl 3-bromo-2-methylbenzoate

  • MDL number MFCD09031772
  • Cas Number : 99548-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3-bromo-o-toluate, 2-Bromo-6-(methoxycarbonyl)toluene
Purity 97
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-3-bromo-2-methylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-bromo-2-methylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart