APO455830799 Apollo Scientific Ltd

4-Bromo-2-iodobenzoic acid

  • MDL number MFCD12767500
  • Cas Number : 1133123-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-186
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-iodobenzoic-acid_OR40655

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-iodobenzoic-acid_OR40655
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart