APO455830800 Apollo Scientific Ltd

2-Bromo-5-iodobenzyl bromide

  • MDL number MFCD15527653
  • Cas Number : 289617-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-(bromomethyl)-4-iodobenzene, alpha,2-Dibromo-5-iodotoluene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55734

Safety Information

signalword Danger
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55734
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart