APO455830803 Apollo Scientific Ltd

2-Bromo-5-iodophenol

  • MDL number MFCD09907878
  • Cas Number : 932372-99-1
  • Molecular Weight : 298.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-iodophenol
Inchi Code 1S/C6H4BrIO/c7-5-2-1-4(8)3-6(5)9/h1-3,9H
Inchi Key VRITXTVQJJMQIT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-iodophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-iodophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart