APO455830806 Apollo Scientific Ltd

2-Bromo-6-methylanisole

  • MDL number MFCD12547891
  • Cas Number : 52200-69-8
  • Molecular Weight : 201.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp -39 to -36
Iupac Name 1-bromo-2-methoxy-3-methylbenzene
density 1.41
Inchi Code 1S/C8H9BrO/c1-6-4-3-5-7(9)8(6)10-2/h3-5H,1-2H3
Inchi Key VUIUOSKXGNEORM-UHFFFAOYSA-N

Description

Synonym 3-Bromo-2-methoxytoluene, 1-Bromo-2-methoxy-3-methylbenzene, 2-Bromo-6-methylphenyl methyl ether
Purity 98%
Density 1.41
Flash Point 62-63/3mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point -39 to -36
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-methylanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-methylanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart