APO455830808 Apollo Scientific Ltd

3-Bromo-5-iodobenzaldehyde

  • MDL number MFCD07779024
  • Cas Number : 188813-09-4
  • Molecular Weight : 310.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-iodobenzaldehyde
Inchi Code 1S/C7H4BrIO/c8-6-1-5(4-10)2-7(9)3-6/h1-4H
Inchi Key XZXJUWDREBPKRI-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-iodobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-iodobenzaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart