APO455830809 Apollo Scientific Ltd

4-Bromo-2-iodobenzaldehyde

  • MDL number MFCD18391619
  • Cas Number : 1261470-87-4
  • Molecular Weight : 310.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2-iodobenzaldehyde
Inchi Code 1S/C7H4BrIO/c8-6-2-1-5(4-10)7(9)3-6/h1-4H
Inchi Key JFFYPBIPKMTEEQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-iodobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-iodobenzaldehyde
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart