APO455830812 Apollo Scientific Ltd

4-Bromo-3-iodoanisole

  • MDL number MFCD00070743
  • Cas Number : 4897-68-1
  • Molecular Weight : 312.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-iodo-4-methoxybenzene
Inchi Code 1S/C7H6BrIO/c1-10-5-2-3-6(8)7(9)4-5/h2-4H,1H3
Inchi Key IGPSHEKLLKFEOJ-UHFFFAOYSA-N

Description

Synonym 1-Bromo-2-iodo-4-methoxybenzene, 4-Bromo-3-iodophenyl methyl ether
Purity 95%
Flash Point 124-126/2mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-iodoanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-iodoanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart