APO455830820 Apollo Scientific Ltd

2-(Cyanomethyl)benzeneboronic acid, pinacol ester

  • MDL number MFCD02179457
  • Cas Number : 325141-71-7
  • Molecular Weight : 243.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Inchi Code 1S/C14H18BNO2/c1-13(2)14(3,4)18-15(17-13)12-8-6-5-7-11(12)9-10-16/h5-8H,9H2,1-4H3
Inchi Key FIARMBRSPSECPP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-cyanomethylbenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2-cyanomethylbenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart