APO455830829 Apollo Scientific Ltd

Isothiazole-3-carboxylic acid

  • MDL number MFCD09834765
  • Cas Number : 4576-90-3
  • Molecular Weight : 129.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 135-137
Iupac Name 3-isothiazolecarboxylic acid
Inchi Code 1S/C4H3NO2S/c6-4(7)3-1-2-8-5-3/h1-2H,(H,6,7)
Inchi Key BMPVWNJQCJQBFW-UHFFFAOYSA-N

Description

Synonym 3-Carboxyisothiazole, 1,2-Thiazole-3-carboxylic acid
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 135-137
Product Link https://store.apolloscientific.co.uk/product/isothiazole-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/isothiazole-3-carboxylic-acid
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart