APO455830833 Apollo Scientific Ltd

3-(N,N-Dipropylaminocarbonyl)benzeneboronic acid

  • MDL number MFCD06659889
  • Cas Number : 850567-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-120
Synonym 3-(Dipropylcarbamoyl)phenylboronic acid
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30302

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30302
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart