APO455830838 Apollo Scientific Ltd

1,3-Thiazole-2-boronic acid, pinacol ester

  • MDL number MFCD08061338
  • Cas Number : 214360-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole, 4,4,5,5-Tetramethyl-2-(1,3-thiazol-2-yl)-1,3,2-dioxaborolane
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47991

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47991
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart