APO455830841 Apollo Scientific Ltd

(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)acetonitrile

  • MDL number MFCD00268751
  • Cas Number : 115615-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2,3-Dihydro-3-oxo-4H-benzo[b][1,4]oxazin-4-yl)acetonitrile, 4-(Cyanomethyl)-3,4-dihydro-3-oxo-2H-1,4-benzoxazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13997

Safety Information

signalword Warning
pcodes P301+P312;P302+P352;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13997
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart