APO455830844 Apollo Scientific Ltd

2,5-Dimethyl-4-methoxybenzeneboronic acid

  • MDL number MFCD06801676
  • Cas Number : 246023-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-190
Synonym 4-Borono-2,5-dimethylanisole, (4-Methoxy-2,5-dimethylphenyl)boronic acid
Purity 96
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/25-dimethyl-4-methoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/25-dimethyl-4-methoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart