APO455830846 Apollo Scientific Ltd

Methyl 4-azaindole-3-carboxylate

  • MDL number MFCD09864655
  • Cas Number : 952800-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 1H-pyrrolo[3,2-b]pyridine-3-carboxylate, 3-(Methoxycarbonyl)-1H-pyrrolo[3,2-b]pyridine
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48619

Safety Information

signalword Warning
pcodes P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48619
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart