APO455830850 Apollo Scientific Ltd

2-Methoxy-5-formylbenzeneboronic acid

  • MDL number MFCD01319044
  • Cas Number : 127972-02-5
  • Molecular Weight : 179.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 160
Iupac Name 5-formyl-2-methoxyphenylboronic acid
Inchi Code 1S/C8H9BO4/c1-13-8-3-2-6(5-10)4-7(8)9(11)12/h2-5,11-12H,1H3
Inchi Key NKKNXLPHCRLBDY-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 160
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-5-formylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-5-formylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart