APO455830852 Apollo Scientific Ltd

5-Bromo-2-methoxy-4-methyl-3-nitropyridine

  • MDL number MFCD06659504
  • Cas Number : 884495-14-1
  • Molecular Weight : 247.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-methyl-3-nitro-2-pyridinyl methyl ether
Inchi Code 1S/C7H7BrN2O3/c1-4-5(8)3-9-7(13-2)6(4)10(11)12/h3H,1-2H3
Inchi Key BGDKJBCVNNWITN-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2-methoxy-3-nitro-4-picoline
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-methoxy-4-methyl-3-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-methoxy-4-methyl-3-nitropyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart