APO455830860 Apollo Scientific Ltd

4-(N-Ethylsulphamoyl)benzeneboronic acid

  • MDL number MFCD07363751
  • Cas Number : 871329-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-136
Synonym 4-Borono-N-ethylbenzenesulphonamide, 4-[(Ethylamino)sulphonyl]benzeneboronic acid
Purity 97
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=30331

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=30331
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart