APO455830861 Apollo Scientific Ltd

2,2-Dimethylnonanoic acid

  • MDL number MFCD02093530
  • Cas Number : 14250-75-0
  • Molecular Weight : 186.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/22-dimethylnonanoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/22-dimethylnonanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart