APO455830867 Apollo Scientific Ltd

Benzyl tert-butyl malonate

  • MDL number MFCD01075175
  • Cas Number : 72594-86-6
  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Benzyl tert-butyl propane-1,3-dioate
Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/benzyl-tert-butyl-malonate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/benzyl-tert-butyl-malonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart