APO455830875 Apollo Scientific Ltd

1-(Pent-1-yl)piperazine

  • MDL number MFCD02093555
  • Cas Number : 50866-75-6
  • Molecular Weight : 156.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-pentylpiperazine
Inchi Code 1S/C9H20N2/c1-2-3-4-7-11-8-5-10-6-9-11/h10H,2-9H2,1H3
Inchi Key MJWWNBHUIIRNDZ-UHFFFAOYSA-N

Description

Flash Point 88-91/20mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-pent-1-ylpiperazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-pent-1-ylpiperazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart