APO455830877 Apollo Scientific Ltd

1-(4-Phenylbut-1-yl)piperazine

  • MDL number MFCD02093540
  • Cas Number : 97480-93-8
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-phenylbutyl)piperazine
Inchi Code 1S/C14H22N2/c1-2-6-14(7-3-1)8-4-5-11-16-12-9-15-10-13-16/h1-3,6-7,15H,4-5,8-13H2
Inchi Key OHFQSNUBVNIZRV-UHFFFAOYSA-N

Description

Synonym 1-Phenyl-4-(piperazin-1-yl)butane
Purity 99%
Flash Point 110-112/0.3mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-phenylbut-1-ylpiperazine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-phenylbut-1-ylpiperazine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart