APO455830879 Apollo Scientific Ltd

4-Nitrophenoxyacetic acid hydrazide

  • MDL number MFCD00051973
  • Cas Number : 75129-74-7
  • Molecular Weight : 211.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 184-186
Iupac Name 2-(4-nitrophenoxy)acetohydrazide
Inchi Code 1S/C8H9N3O4/c9-10-8(12)5-15-7-3-1-6(2-4-7)11(13)14/h1-4H,5,9H2,(H,10,12)
Inchi Key BDPKZWIKLVUGKS-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 184-186
Product Link https://store.apolloscientific.co.uk/product/4-nitrophenoxyacetic-acid-hydrazide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-nitrophenoxyacetic-acid-hydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart