APO455830893 Apollo Scientific Ltd

5-Hydroxybenzo[b]furan

  • MDL number MFCD10699412
  • Cas Number : 13196-10-6
  • Molecular Weight : 134.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzofuran-5-ol
Inchi Code 1S/C8H6O2/c9-7-1-2-8-6(5-7)3-4-10-8/h1-5,9H
Inchi Key IZFOPMSVNDORMZ-UHFFFAOYSA-N

Description

Synonym Benzo[b]furan-5-ol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-hydroxybenzobfuran

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-hydroxybenzobfuran
pictograms GHS07
hstatements H302;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart