APO455830897 Apollo Scientific Ltd

9-Phenyl-9H-carbazole-3-boronic acid

  • MDL number MFCD12196936
  • Cas Number : 854952-58-2
  • Molecular Weight : 287.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-phenyl-9H-carbazol-3-ylboronic acid
Inchi Code 1S/C18H14BNO2/c21-19(22)13-10-11-18-16(12-13)15-8-4-5-9-17(15)20(18)14-6-2-1-3-7-14/h1-12,21-22H
Inchi Key JWJQEUDGBZMPAX-UHFFFAOYSA-N

Description

Synonym 3-Borono-9-phenyl-9H-carbazole
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/9-phenyl-9h-carbazole-3-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/9-phenyl-9h-carbazole-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart