APO455830910 Apollo Scientific Ltd

Methyl 4-bromo-1H-pyrazole-3-carboxylate

  • MDL number MFCD00619124
  • Cas Number : 81190-89-8
  • Molecular Weight : 205.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-bromo-1H-pyrazole-3-carboxylate
Inchi Code 1S/C5H5BrN2O2/c1-10-5(9)4-3(6)2-7-8-4/h2H,1H3,(H,7,8)
Inchi Key KWQDACZQHJCUNK-UHFFFAOYSA-N

Description

Synonym 4-Bromo-3-(methoxycarbonyl)-1H-pyrazole, Methyl 4-bromo-1H-pyrazole-5-carboxylate
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromo-1h-pyrazole-3-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromo-1h-pyrazole-3-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart