APO455830914 Apollo Scientific Ltd

3-Methylthiophene-2-boronic acid

  • MDL number MFCD00236049
  • Cas Number : 177735-09-0
  • Molecular Weight : 141.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 192-196
Iupac Name 3-methyl-2-thienylboronic acid
Inchi Code 1S/C5H7BO2S/c1-4-2-3-9-5(4)6(7)8/h2-3,7-8H,1H3
Inchi Key MOOOGTOEKZWEJA-UHFFFAOYSA-N

Description

Synonym 2-Borono-3-methylthiophene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 192-196
Product Link https://store.apolloscientific.co.uk/product/3-methylthiophene-2-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylthiophene-2-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart