APO455830917 Apollo Scientific Ltd

3-Amino-4-methoxypyridine

  • MDL number MFCD07374959
  • Cas Number : 33631-09-3
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methoxy-3-pyridinamine
Inchi Code 1S/C6H8N2O/c1-9-6-2-3-8-4-5(6)7/h2-4H,7H2,1H3
Inchi Key STWMPIWLSQKHSJ-UHFFFAOYSA-N

Description

Synonym 4-Methoxypyridin-3-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-methoxypyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-methoxypyridine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart