APO455830918 Apollo Scientific Ltd

3-Methylisoquinoline-5-sulphonyl chloride

  • MDL number MFCD11052648
  • Cas Number : 640290-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Chlorosulphonyl)-3-methylisoquinoline, 3-Methyl-2-azanaphthalene-5-sulphonyl chloride
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48043

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=48043
Availability details for
Quantity
  • Availability
  • Add to Cart